Acerca de este artículo
ISM® is a sophisticated safety concept that leaves nothing to chance.
- Intelligent sensor management system (ISM®) internal reference
- Calibration automatically transferred between compatible METTLER TOLEDO meters
- Certificate can be sent to printer or PC
- If the electrode comes with a cable, it will be listed in the order section below
The ISM® technology is compatible with following METTLER TOLEDO Meters: SevenExcellence™, SevenDirect™, SevenGo pro™, SevenGo Duo pro™, and SevenCompact™ (discontinued in June 2022).
The electrode certificate with the factory calibration of an ISM® sensor can be viewed on the meter and transferred to a printer or PC. The last 5 calibration data records are stored in the ISM® sensor and can be viewed at any time, printed out, or transferred to a PC. The maximum temperature to which an ISM® sensor is exposed during a measurement is automatically monitored and can be viewed at any time.
Following the connection of the ISM® sensor, the calibration data of the sensor is transferred to the meter and is used automatically for the subsequent measurement. After calibrating an ISM® sensor, the calibration data is automatically transferred from the meter to the sensor and is stored on the data chip of the sensor.