Orders LinkContactUsLinkComponent
LabLink LED LabLight, PG LifeLink
87" LED LabLight, 3.5W x 2.2H x 221.3L cm (13/8 x 7/8 x 871/8")
  89207-218
 :  LL-87
LabLink LED LabLight, PG LifeLink
  89207-218
LabLink LED LabLight, PG LifeLink
LabLink LED LabLight, PG LifeLink
87" LED LabLight, 3.5W x 2.2H x 221.3L cm (13/8 x 7/8 x 871/8")
$19,848.25
Each (1UN)
The LabLink LabLight is a streamline LED task light for laboratory workbenches
  • Electrical:110-250 VAC 50/60 Hz
  • Efficiency: 75 lumens/watt
  • Power consumption: 2 watts/foot
  • Operating Temperature Range: 32-104ºF (0-40ºC)
  • Illumination: 40 foot candles min. @ 54.6H cm (211/2") (150 lumens/foot); 30 foot candles min. @ 54.6H cm (211/2") for 38.1 cm (15") Units
PG LifeLink’s LabLight is made from 95% recycled materials and Energy Star rated components. The aluminum housing efficiently manages heat from the high power LEDs and stays cool to the touch. The 50,000 hour LED service life makes the LabLight virtually maintenance free. Expected service life is 20 years @ 50 hours/week.
Power connections on both ends of the fixture allow the "daisy chaining" of up to 10’ of light. The LabLight is available in seven models ranging in lengths from 15.125" to 87.125". Add 1.625" to length for each power connection. Length tolerance is ±0.0625".
LabLights are easily attached to metal, plastic or wood furniture using standard magnetic strip mounts or under shelf screw mounts.
The specially designed polycarbonate lens directs up to 95% of the illumination from white LED lamps to the work surface. This feature maximizes light dispersion for uniform task lighting in laboratories and similar applications. Environment is Indoor, Dry Locations.
 : C-UL Listed. ARRA Compliant. ANSI C3.
 : Optional bracket mounts are also available at an additional charge.
  • Description:
    87" LED LabLight
  • Dimensions:
    3.5W x 2.2H x 221.3L cm (13/8 x 7/8 x 871/8")