Eppendorf® UVette® Disposable Cuvettes
Product Details & Documents
These disposable cuvettes allow for measurements in the UV/VIS range with transparency from 230nm to 1600nm
Choose between optical path lengths of 10mm or 2mm with a single 90° rotation. The UVette® is ideally matched for use with the Eppendorf® BioPhotometer® Plus and BioSpectrometer®. For use with other spectrophotometers, the UVette® can be utilized with the aid of an adapter.
Ordering information: The UVette® Routine Pack is supplied in a reclosable box. The UVette® Starter Set includes 80 UVettes and one universal adapter for 8.5mm and 15mm light path heights.
Documents
Eppendorf Photometers and Spectrometers Flyer (CA) Eppendorf Photometers and Spectrometers Flyer (US)
Documentación del producto
Especificaciones
- Description:UVettes, Individually Packaged
- Cat. No.:62111-412
- Environmental certification:Renewable Energy Certificate, ISO 14001
- Environmentally Preferable:Low Manufacturing Impact