Orders LinkContactUsLinkComponent
VWR® Heated Circulating Baths
Supplier:   VWR International
VWR® Heated Circulating Baths
Supplier:  VWR International
Certificates
VWR® Heated Circulating Baths
3 Options Available Below
Product Details & Documents
VWR® Heated Circulating Baths offer precise temperature control, at temperature ranges from ambient +10° to 200°C (depending on the controller).
  • Available in a wide range of sizes
  • Three controller options
  • Ideal for many applications
The large, LCD display on the MX temperature controller is easily viewed across the lab
The Advanced Series Controllers offer Swivel 180™ technology, which allows you to rotate the controller independently from the bath. All heated circulating baths include LidDock™ lid stowing system and DuraTop™, the precision molded phenolic bath top, which remains cooler at high temperatures and resists laboratory chemicals.
The Advanced Digital temperature controller offers a touch-pad, while the Advanced Programmable Temperature Controller includes a full touch-screen display, and programmability (ten 100-step programs).
 : ETL listed. CE marked. MX Controller complies with DIN 12876-1 Class I safety requirements for use with non-flammable liquids. Advanced Digital and Advanced Programmable comply with DIN 12876-1 Class III safety requirements for use with flammable liquids. cETLus compliant.
 : Heated Circulators include reservoir cover; bypass tubing; male inlet and outlet adapters for 3/8, 1/4" and 3/16" (9.5, 6.3 and 4.7 mm ) tubing. 1/4" NPT to M16 adapters also included on 50 Hz models.
 : Every VWR® ISO 17025 NIST Traceable Pre-calibrated Circulating Bath is calibrated and tested by an ISO 17025 accredited lab to ensure the highest standard of accuracy and traceability. Two options are available: Option 1: Standard Pre-calibrated circulating bath is calibrated and tested at both 37°C and 70°C. Option 2: Customer specified pre-calibrated circulating bath provides a choice of 3 different temperature points based on your research requirements. Simply order the stated catalog number and you will receive a protective box containing a work order form allowing you to specify your own calibration temperature points. After completing the work order, you can mail the box using a pre-paid return shipping label or you can scan and email the information to a specified E-mail address. The Avantor calibration team will calibrate the circulating bath based on your instruction.
VWRCATALYST™ can provide an array of services, such as on-site IQ, OQ, PQ, Customized Protocol Generation, Equipment Calibration, Asset Management, and Certification Services (US only). Please call 888-793-2300 or email VWR[email protected] for further details.
Specifications for Product Family
Specifications
  • Catalog No:
  • 89400-974
  • 89202-926
  • 89202-934
  • Max. pressure:
  • 2.3 psi (0.16 bar)
  • 4.3 psi (0.3 bar)
  • 4.3 psi (0.3 bar)
Specifications
Product Family Options
Circulating Baths with MX Controllers
Product Information
  • Temperature Range
    Availability
    Price
  • 89400-974
    15 L Heated Circulating Bath, MX Controller
    15 L Heated Circulating Bath, MX Controller
    Ambient +10 to 135 °C
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $88,761.19
    $88,761.19
    Each (1UN)
       $88,761.19
    Specifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    89400-974
    Max. pressure
    2.3 psi (0.16 bar)
    Product available on GSA Advantage®
Circulating Baths with Advanced Digital Controllers
Product Information
  • Temperature Range
    Availability
    Price
  • 89202-926
    7 L Heated Circulating Bath, Advanced Digital Controller
    7 L Heated Circulating Bath, Advanced Digital Controller
    Ambient +10 to 200 °C
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $95,348.19
    $95,348.19
    Each (1UN)
       $95,348.19
    Specifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    89202-926
    Max. pressure
    4.3 psi (0.3 bar)
    Product available on GSA Advantage®
  • 89202-934
    15 L Heated Circulating Bath, Advanced Digital Controller
    15 L Heated Circulating Bath, Advanced Digital Controller
    Ambient +10 to 200 °C
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $110,004.75
    $110,004.75
    Each (1UN)
       $110,004.75
    Specifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    89202-934
    Max. pressure
    4.3 psi (0.3 bar)
    Product available on GSA Advantage®
Complementary Products
Product Information
  • Catalog #
    Description
    Probe
    Range
    Availability
    Price
  • 89369-150
    Platinum high-accuracy freezer themometer
    Stainless-steel probe with handle
    −100.00...+199.99 °C (−148.00...+391.99 °F)
    Log in for availability
    Each (1UN) - $6,360.63