Nalgene® 50 Platinum-Cured Silicone Tubing for Peristaltic Pumps, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Flexible, durable, and translucent Platinum Silicone tubing is designed to provide extended pump life in peristaltic applications.
- Tubing contains no phthalate plasticizers or peroxides that can leach out into transported fluid
- Autoclavable
This high-purity tubing enables a variety of pump and transfer applications including pharmaceutical, laboratory, bioprocess manufacturing, and food and beverage.
Certifications: Tubing meets FDA, USDA, U.S. Pharmacopeia Class VI, 24th Edition; and 3A sanitary standards.
Ordering information: Supplied in 7.6m (25') coil lengths.
Caution: Note: Not intended for invasive use.
Frequently Bought Together