Ethyl benzoate ≥99%
Glass bottle, 500 g
Catalog # 220123-745
Supplier: SIGMA ALDRICH QUIMICA,S DE RL MX
: E12907-500G
Ethyl benzoate ≥99%
Catalog # 220123-745
Supplier: SIGMA ALDRICH QUIMICA,S DE RL MX
CAS Number: 93-89-0
Jump to:
Product Details & Documents
Sinónimos: Benzoic acid ethyl ester
- CAS Number:93-89-0
- Fórmula:C₉H₁₀O₂
- Número MDL:MFCD00009109
- Molecular Weight:150.18 g/mol
- Índice Merck:13,03802
- Punto de ebullición:415 °C (1013 hPa)
- Punto de fusión:-35 °C
- Punto de fusión:–35 °C
- Densidad:1.05 g/cm³ (20 °C)
- Punto de inflamación:91 °C
- Temperatura de almacenamiento:Ambient
- EINECS:202-284-3
Especificaciones
Especificaciones
- Pack type:Glass bottle
- Size:500 g
Especificaciones
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



