Hexamethylenetetramine ACS
Bottle, 250 g
Catalog # 220063-520
Supplier: SIGMA ALDRICH QUIMICA,S DE RL MX
: 398160-250G
Hexamethylenetetramine ACS
Catalog # 220063-520
Supplier: SIGMA ALDRICH QUIMICA,S DE RL MX
CAS Number: 100-97-0
Jump to:
Product Details & Documents
Sinónimos: Hexamethylenetetramine, Hexamine, Urotropine, Formin
- CAS Number:100-97-0
- Fórmula:C₆H₁₂N₄
- Número MDL:MFCD00006895
- Molecular Weight:140.19 g/mol
- Índice Merck:13,05994
- Punto de fusión:280 °C
- Densidad:1.33 g/cm³ (20 °C)
- Punto de inflamación:250 °C
- Temperatura de almacenamiento:Ambient
- EINECS:202-905-8
- UN:1328
- ADR:4.1,III
Especificaciones
Resultados de prueba de especificación
- Assay:99.0%
Especificaciones
- Pack type:Bottle
- Size:250 g
Especificaciones
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



