Order Entry
Mexico
Orders LinkContactUsLinkComponent
 

Docusate sodium ≥98%

Bottle, 100 g
  220056-112
 
 :  323586-100G
Docusate sodium ≥98%
  220056-112
 :  577-11-7
Docusate sodium ≥98%

Bottle, 100 g

 :  577-11-7
 
$3,667.92
   
Each (1UN)
 
   
 
 :  Dioctyl sulphosuccinate sodium salt, Dioctyl sulphosuccinate, sodium salt, Sodium dioctyl sulphosuccinate, Sodium 1,4-bis((2-ethylhexyl)oxy)-1,4-dioxobutane-2-sulphonate, bis(2-Ethylhexyl) sodium sulphosuccinate
  •  :577-11-7
  •  :C₂₀H₃₇NaO₇S
  •  :MFCD00012455
  •  :444.57 g/mol
  •  :12,03460
  •  :? 180 °C (1013 hPa)
  •  :∼180 °C (1013 hPa)
  •  :~ 155 °C
  •  :∼155 °C
  •  :~ 1.1 g/cm³ (20 °C)
  •  :Ambient
  •  :209-406-4
 

 

  • Assay:
    98%

 

  • Pack type:
    Bottle
  • Size:
    100 g

 

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.