Order Entry
Mexico
ContactUsLinkComponent
Acetonitrilo ≥99.9% CHROMASOLV™ Plus para HPLC
Flammable
Irritant
Acetonitrilo ≥99.9% CHROMASOLV™ Plus para HPLC
Catalog # 220059-671
CAS Number:  
undefined
Flammable
Irritant
Acetonitrilo ≥99.9% CHROMASOLV™ Plus para HPLC
Catalog # 220059-671
Supplier Number:  34998-4L
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Tipo de env.:
    Bottle
  • Tamaño:
    4 L
  • Referencia:
    220059-671
  • Nº de proveedor:
    34998-4L

Specifications

About this item

 :
 :41.05 g/mol
Merck Index:13,00071
Boiling Pt:81.6 °C (1013 hPa)
Boiling Pt:81,6 °C (1013 hPa)
Melting Pt:-45,7 °C
Density:0,782 g/cm³ (20 °C)
Flash Pt:2 °C (closed cup)
Storage Temperature:Ambiente
EINECS:200-835-2
UN:1648
ADR:3,II
REACH:01-2119471307-38
 :GHS02
 :GHS07
CAS Number:75-05-8
Formula:H₃CCN
MDL Number:MFCD00001878