

terc-Butil metil éter ≥99.8% CHROMASOLV™ para HPLC
Catalog # 220059-774
Supplier: SIGMA ALDRICH QUIMICA,S DE RL MX
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Tipo de env.:Glass bottle, narrow mouth
- Tamaño:1 L
- Referencia:220059-774
- Nº de proveedor:34875-1L
Specifications
About this item
:
:88.15 g/mol
Merck Index:13,06059
Boiling Pt:55.2 °C (1013 hPa)
Boiling Pt:55,2 °C (1013 hPa)
Melting Pt:-108,6 °C
Density:0,7405 g/cm³ (20 °C)
Flash Pt:–28 °C
Flash Pt:-28 °C
Storage Temperature:Ambiente
EINECS:216-653-1
UN:2398
ADR:3,II
:GHS02
:GHS07
CAS Number:1634-04-4
Formula:(CH₃)₃COCH₃
MDL Number:MFCD00008812