- Velocidad de flujo:max. 2.0 L/min
- Descripción:PURELAB® flex 2, Water Purification System with Starter Kit, produced water quality: Type I
- Modelo:PURELAB® flex 2
- Inorgánicos @ 25°C:18.2 MΩ.cm
- Detalles eléctricos:100-240V AC, 50-60Hz
- Bacteria:<0.1 CFU/ml with POU filter fitted
- Endotoxin:<0.001 EU/ml with POU Biofilter fitted
- Necesidades diarias promedio:<10 L
- Contenido TOC:<5 ppb
- Referencia:89221-840
- Nº de proveedor:PF2XXXXM1-KIT
The PURELAB® flex range is designed to deliver accuracy, flexibility and ease-of-use. The award-winning system provides perfect water purity for analytical and life science applications which require RO type III water up to ultrapure type I (18.2 MΩ.cm) water. It allows focus on routine test work without concern about the water quality affecting test results.
- Customized settings
- Easy access to the consumables through the front door panels
- Maintenance time is reduced, with less disruption to your work
- Download all of the data to USB for system performance validation
- Clear water purity display for absolute confidence as you dispense
Be in control of your PURELAB® flex by customizing the settings to suit your application
Suggested applications for the flex 2 include mass mpectrometry, molecular biology, electrochemistry, atomic spectroscopy, liquid chromatography, gas chromatography, immunochemistry, spectrophotometry, media / buffer prep, general chemistry.
The flex 1 is designed to dispense water when it is connected to a reservoir or distribution loop. This system works as a dispenser as well as a simple deionization system. The flex 2 includes UV photo-oxidation and TOC monitoring in addition to deionization. The flex 2 features guaranteed water purity with full recirculation through the UV lamp and purification pack right to the point of use for peace of mind. It also features real-time TOC monitoring, which provides complete confidence in organic purity by reducing the level of organics for critical applications.
The starter kit includes the LA652, (pressure regulator), the LC145, (point of use filter, .2 micron), and the LC208, (DI purification cartridge).