VWR® Protective Eyewear with Adjustable Temples
Anti-fog, scratch-resistant, Clear, Black
Supplier: VWR International
Jump to:
Product Details & Documents
Ultra-lightweight nylon frame with soft PVC nose pads.
- Unique ratcheting/adjustable temples lock into place for a custom fit
- Ultra-lightweight nylon frames and soft PVC nose pads to prevent slipping
- 8-base spherical lenses provide excellent side protection
- Anti-fog coating extends lens life
Wrap-around sport styling, designed for men and women features eight base spherical lenses providing excellent side protection. The impact resistant polycarbonate lenses provide 99.9% UV protection and the scratch resistant coating.
: ANSI Z87.1
Especificaciones
- Frame Color:Black
- Lens Coating:Anti-fog, scratch-resistant
- Lens Tint:Clear