Semi-quantitative test strips QUANTOFIX Iron
100 Sticks
Supplier: MACHEREY-NAGEL, INC.
: 91330
Semi-quantitative test strips QUANTOFIX Iron
Catalog # 60787-724
Supplier: MACHEREY-NAGEL, INC.
CAS Number:
Jump to:
Product Details & Documents
For semi-quantitative determinations
Gradation: 0 to 1000mg/L. Color changes from white to dark
Certificates
Buscar por número de loteEspecificaciones
Especificaciones
- Pack type:100 Sticks