Finnpipette® Novus GLP Kits, Thermo Scientific
Proveedor: MOLECULAR BIOPRODUCTS INC MX
Certificados
Acerca de este artículo
Finnpipette® Novus GLP Kits provide ergonomic and easy-to-use Finnpipette Novus pipetters in an all-inclusive kit containing everything you need for pipetting.
Pipettors feature 10 pipetting functions and nine pipetting speeds. Unique to Novus, the adjustable index-finger trigger action prevents common repetitive stress injuries caused by thumb-driven pipettes, and the easy-to-follow prompts guide Novus users through 10 pipetting functions.
Pipettors feature 10 pipetting functions and nine pipetting speeds. Unique to Novus, the adjustable index-finger trigger action prevents common repetitive stress injuries caused by thumb-driven pipettes, and the easy-to-follow prompts guide Novus users through 10 pipetting functions.
Certifications: CE marked.
Ordering information: Includes: two or three Novus pipettes, two or three power supplies, two or three Novus stands, two Finntip Flex racks, Reagent Reservoirs, Finntip tip sample pack, F-series pen, Novus brochure and Pipetting Guide.
Frequently Bought Together