Order Entry
Mexico
ContactUsLinkComponent
Urea en solución acuosa
Urea en solución acuosa
Catalog # 97062-966
Supplier:  VWR
CAS Number:  
undefined
Urea en solución acuosa
Catalog # 97062-966
Supplier:  VWR
Supplier Number:  E720-250ML
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Tamaño:
    250 ml
  • Referencia:
    97062-966
  • Nº de proveedor:
    E720-250ML

Specifications

Specification Test Results

  • Appearance:
    Clear and colorless liquid
  • Urea:
    7.8 Molar - 8.2 Molar

Specification Test Results

About this item

 :
 :60.06 g/mol
Storage Temperature:Ambiente
EINECS:200-315-5
CAS Number:57-13-6
Formula:NH₂CONH₂
MDL Number:MFCD00008022