VWR® READY-PACK™ Ultra Pure Grade TRIS Buffered Saline (TBS)
Catálogo # 97062-370
Proveedor: VWR
Número CAS:
Especificaciones
- Tamaño:2 Packs
- Referencia:97062-370
- Nº de proveedor:0788-2PK
Especificaciones
Acerca de este artículo
Pre-blended TBS buffer
Each Ready-Pack™ prepares 1 liter of a 20X solution of tris buffered saline when dissolved in water. A 1X solution contains 25mM Tris, 140mM Sodium Chloride, and 3.0mM Potassium Chloride.