VWR® TRIS-Acetate-EDTA Buffer, Ultra Pure Grade
Supplier: VWR
Certificates
About this item
TBE is optimized for DNA electrophoresis techniques using both acrylamide and agarose
- High buffering capacity
- High ionic strength
- Permits extended electrophoresis runs without buffer recirculation
- pH ( 1X Concentration) @25°C - 8.1 - 8.5
Each Ready-Pack™ (0478-2PK) prepares 1 L of 10X concentrate.
One 40 L bottle (0478-40L) prepares 4 L of 10X concentrate, or 40 L of 1X solution.
Composition of (1X) TBE Buffer is 0.089M Tris Base; 0.089M Boric Acid; 0.002M EDTA, Free Acid; Final pH 8.3. Made using EDTA, Free Acid. DNase, Protease, RNase free.
Frequently Bought Together