ContactUsLinkComponent
VWR® 25X TRIS-Acetate-EDTA Buffer, Ultra Pure Grade
  97062-384
 :  VWR
 :  
undefined
VWR® 25X TRIS-Acetate-EDTA Buffer, Ultra Pure Grade
  97062-384
 :  VWR
 :  0796-1.6L
 :  
  • Tamaño:
    1.6 L
  • Referencia:
    97062-384
  • Nº de proveedor:
    0796-1.6L
TAE is an extensively used buffer for agarose gel electrophoresis applications requiring high resolution and separation of high molecular weight, double-stranded DNA
TAE is more compatible with in-gel manipulations and band recovery procedures than a TBE buffer. A single strength (1X) solution contains 0.04M Tris-Acetate and 0.001M EDTA with a final pH of 8.0. Low ionic strength. Better resolution of large DNA fragments (>12kbDNA).