Orders LinkContactUsLinkComponent
VWR® MOPS Buffer Solution 10X, Biotechnology Grade
VWR® MOPS Buffer Solution 10X, Biotechnology Grade
500 mL
Catalog # 97062-854
Supplier:   VWR
Número de proveedor #:  E526-500ML
VWR® MOPS Buffer Solution 10X, Biotechnology Grade
Catalog # 97062-854
Supplier:  VWR
CAS Number:  
VWR® MOPS Buffer Solution 10X, Biotechnology Grade
VWR® MOPS Buffer Solution 10X, Biotechnology Grade
500 mL
CAS Number:  
Price
$3,894.68
Log in Para disponibilidad
Each (500ml)
Product Details & Documents
Optimized for RNA Agarose Electrophoresis
Ideal for Use with RNA Formaldehyde Gels.
Especificaciones
Especificaciones
  • Size:
    500 mL