750 Series Carts, Saf-T-Cart
300008-191
- Material:Acero
- Color:Green
- Ancho:19"
- Profundidad:9½" (Max.)
- Altura:47"
- Tipo de asa:U-Handle
- Diámetro de la rueda:10"
- Wheel type:Semi-Pneumatic
- Referencia:300008-191
- Load Capacity:500 lbs.
- Casters:2
- Platform Size:6D×11D"
- Peso:35 lbs.
- Nº de proveedor:339-750-10
- Wheel Width:2¾"
Heavy-duty hand trucks constructed of a tubing with a floor plate. Perfect for cylinder delivery.