ContactUsLinkComponent
Corning® Reusable Plastic Low Form Beakers, PP, Corning
Corning® Reusable Plastic Low Form Beakers, PP, Corning
Catálogo # 89172-050
Proveedor:  Corning
Número de proveedor #: 1000P-600
Product available on GSA Advantage®
undefined
Corning® Reusable Plastic Low Form Beakers, PP, Corning
Catálogo # 89172-050
Proveedor:  Corning
Número de proveedor:  1000P-25
Número de proveedor #: 1000P-600
Product available on GSA Advantage®
Iniciar sesión Para disponibilidad
Select an Option
89 mm (3<sup>1</sup>/<sub>2</sub>")
Select an Option
100 ml
Select an Option
600 ml (20 oz.)
Select an Option
125 mm (5")
Select an Option
89172-050
Select an Option
1000P-600
Acerca de este artículo
PP, translucent.
  • Very good chemical resistance
  • Very good temperature resistance
  • Autoclavable
With printed with blue graduations.
Frequently Bought Together