ContactUsLinkComponent
Corning® Reusable Plastic Graduated Beakers with Handle and Spout, PP, Corning
Corning® Reusable Plastic Graduated Beakers with Handle and Spout, PP, Corning
Catalog # 89172-072
Supplier:  Corning
Supplier #: 1015P-250
Product available on GSA Advantage®
undefined
Corning® Reusable Plastic Graduated Beakers with Handle and Spout, PP, Corning
Catalog # 89172-072
Supplier:  Corning
Supplier Number:  1015P-250
Supplier #: 1015P-250
Product available on GSA Advantage®
Log in for availability
Select an Option
77 mm (3")
Select an Option
5 ml
Select an Option
250 ml (8.5 oz.)
Select an Option
115 mm (4½")
Select an Option
89172-072
Select an Option
1015P-250
About this item
PP, translucent.
  • Smooth surfaces for easy and fast cleaning
  • Printed with blue graduations
  • With handle and spout
Ideal for general laboratory use and a wide variety of applications.
Frequently Bought Together