Orders LinkContactUsLinkComponent
VWR® SSC Buffer
VWR® SSC Buffer
SSC BUFFER, 20X LIQUID CONCENTRATE, 4 L
Catalog # 97062-390
Supplier:   VWR
Número de proveedor #:  0804-4L
VWR® SSC Buffer
Catalog # 97062-390
Supplier:  VWR
CAS Number:  
VWR® SSC Buffer
VWR® SSC Buffer
SSC BUFFER, 20X LIQUID CONCENTRATE, 4 L
CAS Number:  
Price
$2,326.77
Log in Para disponibilidad
Each (4l)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
SSC Buffer is a hybridization buffer for Northern and Southern blots
VWR® offers SSC in various convenient forms: a 20X liquid concentrate, a tablet that prepares 100 ml of a 1X solution, and a Ready-Pack™ that prepares 1 L of 20X concentrate.
Documents
Especificaciones
Especificaciones
  • Description:
    SSC BUFFER, 20X LIQUID CONCENTRATE
  • Size:
    4 L
  • Application:
    Hybridization buffer.
Especificaciones
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 