Orders LinkContactUsLinkComponent
Avantor® ACE® Buffers
Avantor® ACE® Buffers
Supplier:   VWR
Avantor® ACE® Buffers
Supplier:  VWR
Certificates
Avantor® ACE® Buffers
Avantor® ACE® Buffers
1 Options Available Below
Product Details & Documents
These optimized buffers for DNA capillary electrophoresis sequencing provide excellent results, long read lengths, high resolution, and consistent run times
  • Long Read Lengths
  • High Resolution
  • Consistent Run Times
A.C.E.™ 10X Sequencing Buffer concentrate is conveniently diluted to the desired working concentration, thereby providing an easy and cost-effective buffer for your operating systems. VWR® A.C.E.™ 1X Sequencing Buffer comes ready-to-use, no dilution required, offering convenience and time savings. The tinted buffers have a slight pink hue in order to help the user differentiate between VWR® A.C.E.™ buffer and any other water, buffer, or solution in the sequencer. Tinted A.C.E.™ buffers have the same high-quality formulation as all A.C.E.™ products, delivering consistent and reproducible results.
Documents
Certificates
Search by Lot Number
Product Family Options
10x Sequencing Buffers
Product Information