DNA MW Markers, 100 bp Ladder, AMRESCO
Proveedor: VWR
Certificados
Acerca de este artículo
13 fragments ranging from 100 to 3000 bp for easy band identification. Also available pre-mixed with loading dye or with EZ-Vision® One DNA dye.
- Easy band identification
- Available with loading dye
Also available pre-mixed with loading dye or with EZ-Vision® One DNA dye.
Especificaciones
- Environmentally Preferable:Safer Human and Environmental Health
Especificaciones
Información relacionada
Frequently Bought Together