Orders LinkContactUsLinkComponent
Agarose I™, AMRESCO
Agarose I™, AMRESCO
Supplier:   VWR
Agarose I™, AMRESCO
Supplier:  VWR
Certificates
Agarose I™, AMRESCO
Agarose I™, AMRESCO
3 Options Available Below
Product Details & Documents
Agarose I™ is for routine nucleic acid analytical/preparative applications.
  • All-Purpose, High-Purity Agarose
  • Exceptional Band Resolution and Clarity
  • Nuclease- and Protease-free
A standard melting/gelling agarose, suitable for routine nucleic acid analytical/preparative applications. It has a low EEO for shorter electrophoretic runs without compromising resolution. Excellent for blotting techniques and general manipulations.
Product Family Options
Product Information
  • Size
    Availability
    Price
  • 100 g
    Log in for availability
    Log in for availability
    Log in for availability
    Each (100g)
    Each (100g) - $2,114.34
    $2,114.34
    Log in for availability
    Case de 12 (100g)
    Case de 12 (100g) - $2,304.64
    $2,304.64
    Each (100g)
       $2,114.34
    Case de 12 (100g)
       $2,304.64
    Supplier Number: 0710-100G
  • 250 g
    Log in for availability
    Log in for availability
    Log in for availability
    Each (250g)
    Each (250g) - $4,166.14
    $4,166.14
    Log in for availability
    Case de 12 (250g)
    Case de 12 (250g) - $4,541.09
    $4,541.09
    Each (250g)
       $4,166.14
    Case de 12 (250g)
       $4,541.09
    Supplier Number: 0710-250G
  • 500 g
    Log in for availability
    Log in for availability
    Log in for availability
    Each (500g)
    Each (500g) - $5,642.11
    $5,642.11
    Log in for availability
    Case de 12 (500g)
    Case de 12 (500g) - $6,149.90
    $6,149.90
    Each (500g)
       $5,642.11
    Case de 12 (500g)
       $6,149.90
    Supplier Number: 0710-500G