Orders LinkContactUsLinkComponent
VWR® pH Standards
Supplier:   VWR
VWR® pH Standards
Supplier:  VWR
Certificates
VWR® pH Standards
VWR® pH Standards
4 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
 : The pH STANDARD kit includes 500 mL of pH 4.0, pH 7.0 and pH 10.0 standards.
Product Family Options
Información del producto
  • Size
    pH
    Color
    Availability
    Price
  • 500 mL
    10.0
    Blue
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (500ml)
    Each (500ml) - $1,962.52
    $1,962.52
    Each (500ml)
       $1,962.52
    Supplier Number: E464-500ML
  • Kit
    4.0/7.0/10.0
    Multiple
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $4,763.50
    $4,763.50
    Each (1UN)
       $4,763.50
    Supplier Number: E603-KIT
  • 500 mL
    7.0
    Yellow
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (500ml)
    Each (500ml) - $1,369.21
    $1,369.21
    Each (500ml)
       $1,369.21
    Supplier Number: E459-500ML
  • 500 mL
    4.00
    Pink
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (500ml)
    Each (500ml) - $1,729.71
    $1,729.71
    Each (500ml)
       $1,729.71
    Supplier Number: E452-500ML
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.