Orders LinkContactUsLinkComponent
VWR® TRIS EDTA Buffer, Biotechnology Grade
VWR® TRIS EDTA Buffer, Biotechnology Grade
Supplier:   VWR
VWR® TRIS EDTA Buffer, Biotechnology Grade
Supplier:  VWR
Certificates
VWR® TRIS EDTA Buffer, Biotechnology Grade
VWR® TRIS EDTA Buffer, Biotechnology Grade
2 Options Available Below
Product Details & Documents
Product Family Options
Product Information
  • Size
    Availability
    Price
  • 100 mL
    Log in for availability
    Log in for availability
    Each (100ml)
    Each (100ml) - $920.74
    $920.74
    Each (100ml)
       $920.74
    Supplier Number: E112-100ML
  • 500 mL
    Log in for availability
    Log in for availability
    Each (500ml)
    Each (500ml) - $1,482.97
    $1,482.97
    Each (500ml)
       $1,482.97
    Supplier Number: E112-500ML