HistoChoice® Clearing Agent, AMRESCO
HistoChoice® Clearing Agent, 4L
Supplier: VWR
Supplier #: H103-4L
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Less hazardous replacement for xylene and toluene. Compatible with organic mounting media, clearing agent is designed to work with isopropanol in automated processors. It also offers faster wax dissolution and slower evaporation than xylene with no oily residue. Clearing agent can be recycled by distillation.
- Less toxic replacement for xylene
- Virtually odorless
- Faster wax dissolution and slower evaporation than xylene
Documents
Specifications
Specifications
- Description:HistoChoice® Clearing Agent
- Size:4L