Orders LinkContactUsLinkComponent
VWR® Yeast Extract, Bacteriological Grade
Supplier:   VWR
VWR® Yeast Extract, Bacteriological Grade
Supplier:  VWR
Certificates
VWR® Yeast Extract, Bacteriological Grade
VWR® Yeast Extract, Bacteriological Grade
3 Options Available Below
Product Details & Documents
High-purity extract used for media supplements.
  • High-purity extract
  • Contains Vitamin B complex
  • Used for media supplements
High-purity extract used for media supplements. Extract is the water soluble portion of autolyzed yeast, containing a source of Vitamin B complex.
Product Family Options
Información del producto
  • Size
    Availability
    Price
  • 100 g
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (100g)
    Each (100g) - $2,142.38
    $2,142.38
    Log in Para disponibilidad
    Case de 12 (100g)
    Case de 12 (100g) - $2,335.20
    $2,335.20
    Each (100g)
       $2,142.38
    Case de 12 (100g)
       $2,335.20
    Supplier Number: J850-100G
  • 500 g
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (500g)
    Each (500g) - $2,181.92
    $2,181.92
    Log in Para disponibilidad
    Case de 4 (500g)
    Case de 4 (500g) - $2,378.29
    $2,378.29
    Each (500g)
       $2,181.92
    Case de 4 (500g)
       $2,378.29
    Supplier Number: J850-500G
  • 1 kg
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1kg)
    Each (1kg) - $2,665.63
    $2,665.63
    Log in Para disponibilidad
    Case de 4 (1kg)
    Case de 4 (1kg) - $2,905.54
    $2,905.54
    Each (1kg)
       $2,665.63
    Case de 4 (1kg)
       $2,905.54
    Supplier Number: J850-1KG