Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Tipo de env.:Bottle, amber
- CAS No.:9003-99-0
- Tamaño:25000 units
- Condiciones de almacenamiento:Freezer
- MDL No.:MFCD00071339
- Nombre de la enzima:Peroxidase
- Tested applications:for biochemistry
- Referencia:97061-456
- Nº de proveedor:0343-25,000U
HRP catalyzes the decomposition of hydrogen peroxide in solution causing the oxidation of a number of substrates.
Used extensively as a label for immunohistological assays and ELISA. Its exceptional stability in powdered and solution form enhances its desirability as an enzyme conjugate.