Allsop® MousePad Pro™ Memory Foam Mouse Pad, Essendant LLC MS
500041-542
- Color:Black
- Altura:1"
- Environmentally Preferable:Sustainable Materials
- Referencia:500041-542
- Profundidad:8¼"
- Anchura:7¼"
- Nº de proveedor:ASP30203
Designed to relieve stress on pressure points while using your mouse, this memory foam mouse pad conforms and adjusts to the contours of your wrist.
Does not contain petroleum-based materials, such as EVA, PVC or PU. Soft-cloth surface is attached to the non-skid base without glue or harmful chemicals. Non-skid Eco-base™ is made of 60% natural rubber and 40% post-industrial recycled material.