Scotch® Transparent Glossy Tape, Essendant LLC MS
Catalog # 500043-733
Supplier: ESSENDANT LLC FKA LAGASSE
Specifications
- Color:Clear
- Tamaño:3/4" x 1000"
- Referencia:500043-733
- Core Size:25.4 mm (1")
- Se ajuste:Glossy
- Nº de proveedor:MMM600K24
Specifications
About this item
Offices and homes need supplies that can handle a wide range of jobs--making this tape an ideal addition
The excellent holding power and instant adhesion means you can use this tape for a wide range of applications. The long aging, stain- and moisture-resistant makeup is ideal for label protection and multi-purpose sealing. The tape offers you a glossy, high-clarity finish that looks great. Plus, it's photo safe. The tape is designed for use in desktop dispensers, allowing for a quick and easy application.