Scienceware® Poxygrid® Bench-Top Biohazard Bag Holder Kit, Bel-Art
Catalog # 11215-857
Supplier: Bel-Art Products, a Part of SP
Specifications
- Capacidad:1,6 L (0.43 gal.)
- Descripción:Biohazard bags
- Grosor:0.018 mm (0.72 mil.)
- Referencia:11215-857
- An x La:220×280 mm (8.5×11")
- Nº de proveedor:F13166-0000
Specifications
About this item
Economical and convenient, this benchtop disposal system is perfect for collecting pipette tips, microcentrifuge tubes and more during daily use. The rectangular steel wire frame holds a polyethylene bag 22 x 28 cm (8¹/₂ x 11) and is coated in easy to clean, durable, orange color epoxy. The snug fitting polypropylene plastic cover helps hold the bag securely in place. It is reusable, can be cleaned by hand or in automated dish washers, and autoclaved as needed.
- Use as a biohazard material handler: Small size allows the bag to be inverted over your hand to safely pick up waste
- Steam autoclavable at 121 °C (250 °F)
For a space-saving disposal unit in fume hoods or on bench tops, use with Poxygrid® Benchtop Biohazard Bag Holder (11215-508) and Cover (89237-571).