Orders LinkContactUsLinkComponent
Tool Mounting Bar, Perfex
Tool Mounting Bar, Perfex
Supplier:   PERFEX CORP
Tool Mounting Bar, Perfex
Supplier:  PERFEX CORP
Certificates
Tool Mounting Bar, Perfex
Tool Mounting Bar, Perfex
2 Options Available Below
Product Details & Documents
Holds three tools in an 18" friction-fit channel, allowing instant repositioning along the bar length.
  • Individual mount is also available
Securely holds a wide range of tools, up to 11/2" diameter. Mounting holes are 16" on center for convenient attachment to studs, drywall or other wall surfaces.
Product Family Options
Información del producto
  • Color
    Availability
    Price
  • 22940-072
    Tool Mounting Bar
    Tool Mounting Bar
    White
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,121.98
    $1,121.98
    Each (1UN)
       $1,121.98
    Supplier Number: 4000
  • 22940-074
    Individual Tool Mount
    Individual Tool Mount
    Black
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $225.16
    $225.16
    Each (1UN)
       $225.16
    Supplier Number: 4001