VWR® Maximum Coveralls
These are the cleanest and most durable coveralls available within the VWR® Protection line. Manufactured from a specially formulated breathable microporous fabric, these garments are distinguished by their exceptional comfort benefits and superior barrier protection capabilities.
- Superior fluid and particulate barrier
- Lowest level of particle shedding
- Light chemical-splash resistance
- Breathable to ensure user comfort
- Soft, cloth-like fabric with maximum tear strength
- Bonded seam option available
The coveralls provide disposable barrier protection against fluids, biologicals, particulates, and light chemical splash.
Coveralls are rigorously tested and manufactured in an ISO Certified facility under stringent process controls to ensure that each garment meets exacting quality standards and performs to specification. Lot controlled and validated through independent laboratory testing.
All coveralls feature inset sleeves, front zipper, and elastic cuffs, ankles, and back. Also available with or without an attached elastic hood. Garments equipped with attached boots have a long-wear anti-skid sole. Serged seams are standard on all coveralls unless otherwise noted (bonded seams also available).
A high vapor transmission rate keeps the wearer comfortable, even throughout extended use. Appropriate for industrial, pharmaceutical, food processing, construction, and certain environmental cleanup applications.
: Tested and manufactured in an ISO certified facility under stringent process controls to ensure that each product meets exacting quality standards and performs to specification. Products are validated through independent laboratory testing.
: 25/case.
These are the cleanest and most durable coveralls available within the VWR® Protection line. Manufactured from a specially formulated breathable microporous fabric, these garments are distinguished by their exceptional comfort benefits and superior barrier protection capabilities.
- Superior fluid and particulate barrier
- Lowest level of particle shedding
- Light chemical-splash resistance
- Breathable to ensure user comfort
- Soft, cloth-like fabric with maximum tear strength
- Bonded seam option available
The coveralls provide disposable barrier protection against fluids, biologicals, particulates, and light chemical splash.
Coveralls are rigorously tested and manufactured in an ISO Certified facility under stringent process controls to ensure that each garment meets exacting quality standards and performs to specification. Lot controlled and validated through independent laboratory testing.
All coveralls feature inset sleeves, front zipper, and elastic cuffs, ankles, and back. Also available with or without an attached elastic hood. Garments equipped with attached boots have a long-wear anti-skid sole. Serged seams are standard on all coveralls unless otherwise noted (bonded seams also available).
A high vapor transmission rate keeps the wearer comfortable, even throughout extended use. Appropriate for industrial, pharmaceutical, food processing, construction, and certain environmental cleanup applications.
: Tested and manufactured in an ISO certified facility under stringent process controls to ensure that each product meets exacting quality standards and performs to specification. Products are validated through independent laboratory testing.
: 25/case.
- Catalog No:
- 414004-308
- 414004-320
- Fabric:
- Cloth Like Polypropylene
- Cloth Like Polypropylene
- Closure type:
- Zip closure
- Zip closure
- Cleanroom class:
- 100/ISO 5
- 100/ISO 5
- Pockets:
- Without pockets
- Without pockets
- Gender:
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Garment application:
- Cleanroom
- Cleanroom
- Air permeability:
- 0.06 cu. ft./min./sq. ft.
- 0.06 cu. ft./min./sq. ft.
- Bacterial filtration efficiency:
- >99.9%
- >99.9%
- Particle shedding (Helmke Drum):
- Level I
- Level I
- Water resistance:
- Zero Penetration
- Zero Penetration
- H₂O vapor transmission rate (avg.):
- 777 g/m²/24 h
- 777 g/m²/24 h
- Tensile strength:
- 25.2 lbs./in (warp); 29.4 lbs./in (filling)
- 25.2 lbs./in (warp); 29.4 lbs./in (filling)
- Tearing strength:
- 4.6 lbs./in (lengthwise yarns); 5.7 lbs./in (widthwise yarns)
- 4.6 lbs./in (lengthwise yarns); 5.7 lbs./in (widthwise yarns)
- Thickness:
- 0.22 mm
- 0.22 mm
- Weight:
- 63 g/m²
- 63 g/m²
Product Family Options
Coveralls with attached hood and boots
- ColorClothing SizeSleevePackaging
- WhiteL34"25/caseCase (25UN)$1,820.87Paleta de 24 (25UN)$1,984.74
Specifications
Cat. No.414004-308FabricCloth Like PolypropyleneClosure typeZip closureCleanroom class100/ISO 5PocketsWithout pocketsGenderUnisexSleeve or leg lengthLongGarment applicationCleanroomAir permeability0.06 cu. ft./min./sq. ft.Bacterial filtration efficiency>99.9%Particle shedding (Helmke Drum)Level IWater resistanceZero PenetrationH₂O vapor transmission rate (avg.)777 g/m²/24 hTensile strength25.2 lbs./in (warp); 29.4 lbs./in (filling)Tearing strength4.6 lbs./in (lengthwise yarns); 5.7 lbs./in (widthwise yarns)Thickness0.22 mmWeight63 g/m²
Coveralls with attached boots
- ColorClothing SizeSleevePackaging
- WhiteM34"25/caseCase (25UN)$2,249.22Paleta de 24 (25UN)$2,451.64
Specifications
Cat. No.414004-320FabricCloth Like PolypropyleneClosure typeZip closureCleanroom class100/ISO 5PocketsWithout pocketsGenderUnisexSleeve or leg lengthLongGarment applicationCleanroomAir permeability0.06 cu. ft./min./sq. ft.Bacterial filtration efficiency>99.9%Particle shedding (Helmke Drum)Level IWater resistanceZero PenetrationH₂O vapor transmission rate (avg.)777 g/m²/24 hTensile strength25.2 lbs./in (warp); 29.4 lbs./in (filling)Tearing strength4.6 lbs./in (lengthwise yarns); 5.7 lbs./in (widthwise yarns)Thickness0.22 mmWeight63 g/m²