Puritan® Double Cotton Tipped Applicator, Wood Handle, Puritan Medical Products
89133-810
Product available on GSA Advantage®
- Referencia:89133-810
- Longitud del mango:147.6 mm (5.812")
- Head Width:4.78 mm (0.188")
- Head Length:15.9 mm (0.625")
- Handle Width:2.11 mm (0.083")
- Nº de proveedor:821-WC DBL
For precision applications such as jewelry manufacturing or art restoration
Double-ended cotton head for dual use; one tapered end and one regular end. Rigid wood handle. Nonsterile. Made in USA.
: Packaged 500 per box (packed as 5 bags of 100), 10 boxes per case.