Puritan® Swab, Cotton Tip, Puritan Medical Products
Catalog # 10805-142
Supplier: PURITAN MEDICAL PRODUCTS COMP
Product available on GSA Advantage®
Specifications
- Esterilidad:Non sterile
- Referencia:10805-142
- Longitud del mango:76.2 mm (3.000")
- Head Width:4.800 mm (0.188")
- Head Length:15.9 mm (0.625")
- Handle Width:2.50 mm (0.098")
- Nº de proveedor:803-WC
Specifications
About this item
All-purpose cleaning and applicator swab.
Standard and large cotton tip options available. Rigid northern white birch shaft.