VWR® Irradiated Coveralls
Supplier: VWR International
Certificates
About this item
Irradiated coveralls are manufactured from a specially formulated breathable microporous fabric that provides significant fluid and barrier protection.
- Excellent fluid and particulate barrier
- Extremely low level of particle shedding
- Superior breathability and water vapor transmission rate
- Soft, cloth-like fabric
Garments exhibit excellent water vapor tranmission to optimize user comfort. Coveralls are gamma irradiated to a Sterility Assurance Level (SAL) of 10-4 to address the concern of bio-burden contamination. Manufactured and packaged in an ISO Class 8 (FED-STD-209E Class 100,000/M3.5) cleanroom.
Certifications: Coveralls are rigorously tested and manufactured in an ISO Certified facility under stringent process controls to ensure that each product meets exacting quality standards and performs to specification. Products are validated through independent lab testing.
Ordering information: Packaged individually in sealed polyethylene bags contained within a larger carton liner. Each case is supplied with a certificate of irradiation.
Specifications
- Catalog No:
- 414004-450
- 414004-451
- Aplicaciones de vestimenta:
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Permeabilidad al aire:
- 0.06
- 0.06
- Eficiencia de filtración bacteriana:
- 0.999
- 0.999
- Difusión de partículas (Helmke Drum):
- Level I
- Level I
- Water resistance:
- Zero Penetration
- Zero Penetration
- Tasa de transmisión de vapor de H₂O (avg.):
- 777 g/m²/24 hrs.
- 777 g/m²/24 hrs.
- Velocidad de transmisión de vapor de agua:
- 777
- 777
- Resistencia a la tracción:
- 21 (Filling)
- 21 (Filling)
- Fuerza desgarradora:
- 8.7 (Widthwise Yarns)
- 8.7 (Widthwise Yarns)
- Sint. Resistencia a la penetración de sangre:
- Pass
- Pass
- Grosor:
- 0.21 mm
- 0.21 mm
- Peso:
- 56 g/m²
- 56 g/m²
Specifications
Related Information
Certificates
Search by Lot Number414004-449_P 414004-450_P 414004-451_P 414004-452_P 414004-453_P
BSE/TSE Certificates
More
Frequently Bought Together