ContactUsLinkComponent
VWR® Maximum Protection Antifog Full Face Shields
VWR® Maximum Protection Antifog Full Face Shields
Catálogo # 414004-679
Proveedor:  VWR International
undefined
VWR® Maximum Protection Antifog Full Face Shields
Catálogo # 414004-679
Proveedor:  VWR International
Especificaciones
  • Descripción:
    Elastic headstrap
  • Dimensiones:
    33.5L×19H cm (13³/₁₆×7¹/₂")
  • Grosor:
    0.18 mm, ±0.02
  • Referencia:
    414004-679
Especificaciones
Acerca de este artículo
VWR® maximum protection antifog full face shields provide total facial splash protection, and are available with a choice of headband styles to ensure security and comfort throughout extended use.
  • Protect front and side of face from fluid splash
  • Low-glare shield provides distortion-free vision
  • Latex-free
Face shields are rigorously tested and manufactured in an ISO Certified facility under stringent process controls to ensure that each product meets exacting quality standards. Products are validated through independent lab testing.
Face shields feature an antifog coating and fit easily over eyewear. A soft, polyurethane foam sponge fits snugly against the forehead. Equipped with elastic or Velcro® headband for a secure fit.
Embalaje: Packaged 10 face shields per pack, 5 packs per box, 4 boxes per case.