Orders LinkContactUsLinkComponent
Tax-Fre® Dry Mops, Contec®
Tax-Fre® Dry Mops, Contec®
Tax-Fre® Dry Mops, Contec®
Supplier:  CONTEC, INC. MX
Certificates
Tax-Fre® Dry Mops, Contec®
Tax-Fre® Dry Mops, Contec®
2 Options Available Below
Product Details & Documents
Designed for dry mopping floors and walls, the Tax-Fre® Dry Mop utilizes the same low-residue material as Tax-Fre® wipes.
  • Utilizing a low-residue pressure-sensitive material, this dry floor mop provides superior pick-up in areas where wet mopping is not an option
  • Will not scratch or damage sensitive surfaces
  • Dramatically reduces clean-up time
  • Ideal for use when liquids are not recommended
The 23×5" (58.4×12.7 cm) trapezoid-shaped swivel head attached to a lightweight 5' (1.5 m) handle enables the operator to clean in hard to each areas. All hardware is cleanroom compatible. The unique construction of the Tax-Fre® mop head allows the use of multiple sheets that can easily be removed when contaminated, revealing a fresh sheet for continued cleaning.
Product Family Options
Product Information
  • Dimensions
    Packed
    Availability
    Price
  • 46610-130
    Tax-Fre® Dry Mop Swivel Frame
    Tax-Fre® Dry Mop Swivel Frame
    58.4×12.7×3 cm (23×5×1.25")
    1/Case
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $2,141.18
    $2,141.18
    Each (1UN)
       $2,141.18
    Supplier Number: UF211
  • 12777-950
    Tax-Fre® Dry Mop
    Tax-Fre® Dry Mop
    30.5×71 cm (12×28")
    50/Bag, 4 Bags/Case
    Log in for availability
    Log in for availability
    Case (200UN)
    Case (200UN) - $7,038.20
    $7,038.20
    Case (200UN)
       $7,038.20
    Supplier Number: TF-1228
  • 94052-810
    Tax-Fre® Tack Cloth, Quarterfolded
    Tax-Fre® Tack Cloth, Quarterfolded
    25.4×43.2 cm (10×17")
    50/Bag, 8 Bags/Case
    Discontinued
    Supplier Number: -