Orders LinkContactUsLinkComponent
CONSTIX®, Sealed Polyester Swabs, SP-3 Series, Contec®
CONSTIX®, Sealed Polyester Swabs, SP-3 Series, Contec®
Sealed Polyester Swabs, SP-3 Series, 125 mm (4.9"), 3 mm (0.12"), 27.4 mm (1.08"), 9.9 mm (0.39"), 152.4 mm (6")
Catalog # 68310-282
Supplier:   CONTEC, INC. MX
Número de proveedor #:  SP-3
CONSTIX®, Sealed Polyester Swabs, SP-3 Series, Contec®
Catalog # 68310-282
Supplier:  CONTEC, INC. MX
CONSTIX®, Sealed Polyester Swabs, SP-3 Series, Contec®
CONSTIX®, Sealed Polyester Swabs, SP-3 Series, Contec®
Sealed Polyester Swabs, SP-3 Series, 125 mm (4.9"), 3 mm (0.12"), 27.4 mm (1.08"), 9.9 mm (0.39"), 152.4 mm (6")
Price
$871.59
Log in Para disponibilidad
Case (50UN)
Product Details & Documents
The CONTIX® SP-3 is a sealed polyester swab consisting of 100% knit polyester fabric thermally bonded to a polypropylene handle.
  • Polyester head features extremely low particulate and residue levels, providing lowest risk of contamination
  • Polypropylene handle provides excellent resistance to most common solvents, acids, and bases
The head of the swab is thermally attached to the handle without adhesives or binders, and is made from cleanroom laundered polyester fabric. The SP-3 swab is durable, absorbent, and features extremely low particulate and residue levels.
Documents
Especificaciones
  • Description:
    Sealed Polyester Swabs, SP-3 Series
  • Handle Length:
    125 mm (4.9")
  • Overall Length:
    152.4 mm (6")
  • Head Width:
    9.9 mm (0.39")
  • Head Length:
    27.4 mm (1.08")
  • Handle Width:
    3 mm (0.12")