CONSTIX®, Sealed Foam Swabs, SF-13 Series, Contec®
Catalog # 68310-254
Supplier: CONTEC, INC. MX
Small Business Enterprise
Specifications
- Descripción:Sealed Foam Swabs, SF-13 Series
- Referencia:68310-254
- Longitud del mango:63.5 mm (2.5")
- Longitud total:81.3 mm (3.2")
- Head Width:0.76 mm (0.03")
- Head Length:17.8 mm (0.7")
- Handle Width:2.5 mm (0.10")
- Nº de proveedor:SF-13
Specifications
About this item
CONSTIX® SF-13 swabs consist of 100ppi open-cell foam tightly sealed around the tip of a glass-filled polypropylene handle. The foam is secured to the handle without the use of adhesives or binders.
- Polyurethane foam head provides good particle entrapment and removal
The foam head conforms tightly to the pointed shape of the swab handle to provide a very fine tip that enables access to small areas and crevices. The polyurethane foam head provides good particle entrapment and removal and is very low in particles and fibers.
The absorbent foam enables solvents, such as isopropanol and ethanol, to be readily sorbed for applications involving solvent delivery or removal. Though compatible with many common solvents, the SF-13 swab is not recommended for use with ketones such as acetone or MEK.
Ordering information: Laundered and double bagged.