Orders LinkContactUsLinkComponent
STERI-Buffer®, Veltek Associates
STERI-Buffer®, Veltek Associates
STERI-Buffer®, Veltek Associates
Certificates
STERI-Buffer®, Veltek Associates
STERI-Buffer®, Veltek Associates
2 Options Available Below
Product Details & Documents
Developed for use as a buffer solution and diluent in lab testing such a bioburden testing, pharmaceutical microbial limits testing, and AET testing and as a rinsing agent."
Sealed with a “no tamper” strip. Filtered at 0.2µm. Gamma-irradiated, double-bagged, and delivered with lot-specific sterile documentation.
Documents
Product Family Options
Información del producto
  • Size
    Availability
    Price
  • 14222-664
    Phosphate buffer solution
    Phosphate buffer solution
    90 mL (3.0 oz.)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (72UN)
    Case (72UN) - $13,156.52
    $13,156.52
    Case (72UN)
       $13,156.52
    Supplier Number: SB100-90
  • 14222-662
    Phosphate buffer solution
    Phosphate buffer solution
    99 mL (3.3 oz.)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (72UN)
    Case (72UN) - $12,752.03
    $12,752.03
    Case (72UN)
       $12,752.03
    Supplier Number: SB100-99