ContactUsLinkComponent
STERI-PEROX, Veltek Associates
STERI-PEROX, Veltek Associates
Catalog # 40300-604
undefined
STERI-PEROX, Veltek Associates
Catalog # 40300-604
Supplier Number:  SPER-02-3%
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Descripción:
    STERI-PEROX®
  • Tamaño:
    3.8 L (1 gal.)
  • Referencia:
    40300-604
  • Esterilidad:
    Sterile
  • Concentración:
    3%
  • Nº de proveedor:
    SPER-02-3%
Specifications
About this item
Sterile, ready-to-use hydrogen peroxide solution for pharmaceutical and biotechnology operations. Formulated with USP Water for Injection
  • Filled in an ISO 5 (Grade A/B, Former Class 100)
  • Prefiltered to 0,2 um
  • Completely traceable and validated for sterility and shelf life
  • Avaliable in 3% and 6% concentrations
  • 16 oz and 32 oz containers come with sterile spray attachments
  • Low residue content
STERI-PEROX reduces exposure concerns for VOC’s in cleanroom operations, leaves a low remaining residue, and is designed for most washable, non-porous, hard, inanimate environmental surfaces.
Certifications: Delivered with lot specific Certificate of Analysis and Certificate of Sterility.
Packaging: The products are packaged to comply with the requirements for transportation into a classified area. Each container is double bagged packaged sterile.