MicroSeal® VP Sealed Edge Polyester Knit Wipers, Berkshire
Wipers are made from 100% continuous filament knit polyester.
- Lightweight, tight-knit construction resists abrasion, minimizes picking and snagging while cleaning rough surfaces
- Cleanroom laundered wipers have ultrasonically sealed edges
Ideal for ISO Class 3 (FED-STD-209E Class 1/M1.5) and less critical environments. Works well with isopropyl alcohol, acetone, and other solvents. Suitable for cleaning areas with oxidation deposits, photolithography process areas, flat panels, and optics. Steam autoclavable for aseptic environments. Laundered and packaged in an ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom.
: Complies with RoHS and REACH requirements.
- Catalog No:
- 94000-970
- Peso base:
- 133 g/m²
- Absorbancia, extrínseca:
- 442 ml/m²
- Absorbancia, intrínseca:
- 3.3 ml/g
- Absorbancia, velocidad:
- 1 sec.
- Extraíbles:
- 0.0024% using deionized water, 0.011% using isopropyl alcohol
- Fibras (>100 µm):
- 0.038/cm²
- Particle generation (>=0,5 µm):
- 0.55 x 10³/m²
- Iones - calcio:
- 0.02 ppm
- Iones - magnesio:
- 0.0095 ppm
- Iones - sodio:
- 0.035 ppm
Frequently Bought Together