VWR® Nonwoven Wipers
414004-514
- Descripción:Knife cut edges
- Matriz:Cellulose 55%/polyester 45%
- Extraíbles:<0.03 g/m² using deionized water,
<0.02 g/m² using isopropyl alcohol - Peso base:68 ±3 g/m²
- Particle generation (>=0,5 µm):98×10⁶/m²
- Absorbancia, extrínseca:308 ml/m²
- Absorbancia, velocidad:<1 s
- Absorbancia, intrínseca:4.7 ml/g
- Iones - sodio:<47 ppm
- Iones, cloruro:<13 ppm
- Referencia:414004-514
- Empaquetado:150/bag
- An x La:30.5×30.5 cm (12×12")
Made from a hydroentangled blend that combines the strength and cleanliness of synthetic polyester with the high absorbency of cellulose. These soft, economical wipers are designed for cleaning surfaces and controlling spills in ISO Class 6 (FED-STD-209E Class 1000/M4.5) and less critical environments. Blend contains 45% polyester and 55% cellulose. No chemical binders. Knife-cut edges. Color: white.
- Manufactured and packed in cleanroom environment
- Free from silicon, amides, and DOP contamination
- Low lint and satisfactory softness to the touch
- High absorbency
- Tensile strength
- Soft edges
These soft, economical wipers are designed for cleaning surfaces and controlling spills in ISO Class 6 (FED-STD-209E Class 1000/M4.5) and less critical environments.
: Wipers comply with RoHS 2002/95/EC.
: Wipers are vacuum-packed in solvent-safe, cleanroom-compatible polyethylene with a label on each inner bag.