Orders LinkContactUsLinkComponent
M-Zone MegaSorb™ Microfiber String Mops, Micronova
M-Zone MegaSorb™ Microfiber String Mops, Micronova
   Opciones sostenibles disponibles
Supplier:   MICRONOVA MFG INC
M-Zone MegaSorb™ Microfiber String Mops, Micronova
Supplier:  MICRONOVA MFG INC
   Opciones sostenibles disponibles
Certificates
M-Zone MegaSorb™ Microfiber String Mops, Micronova
2 Options Available Below
Product Details & Documents
A highly absorbent mop that combines the cleaning efficiency of microfiber with the ease of a looped string mop. The tufted strands are comprised of polyester and nylon filaments, designed to pick up liquids and tiny particles in controlled and critical environments.
  • Highly absorbent and low-linting
  • Polyester and nylon tufted microfiber tubes
  • Picks up tiny particles where cotton fibers just roll over
  • White microfiber option includes an additional scrubbing band
  • Use with MH-1 handle or QDMH-1 MicroMop adapter with SSU series handles
  • Compatible with alcohol and strong disinfectants
  • Launderable for reuse
  • Autoclavable
The M-Zone™ MegaSorb™ Microfiber String Mop features distinctive microfiber tubes with tufted strands made of polyester and nylon filaments, 30 times smaller than cotton fibers, that work to pick up tiny particles.
Specifications for Product Family
Especificaciones
  • Catalog No:
  • 89092-124
  • 89140-706
  • Environmentally Preferable:
  • Waste Reducing
  • Waste Reducing
Especificaciones
Product Family Options
Mop Heads
Información del producto
  • Material
    Color
    Availability
    Price
  • 89092-124
    MegaSorb™ mop head, 12 oz.
    MegaSorb™ mop head, 12 oz.
    Microfiber
    White
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (12UN)
    Case (12UN) - $9,470.00
    $9,470.00
    Case (12UN)
       $9,470.00
    Supplier Number: MZCRMS11-12
    Especificaciones:
    Minority or Woman-Owned Business Enterprise
    Small Business Enterprise
      Waste Reducing
    More
    Specifications
    Cat. No.
    89092-124
    Environmentally Preferable
    Waste Reducing
    Minority or Woman-Owned Business Enterprise
    Small Business Enterprise
      Waste Reducing
  • 89140-706
    MegaSorb™ mop head, 10 oz., bulk pack
    MegaSorb™ mop head, 10 oz., bulk pack
    Microfiber
    Blue
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (50UN)
    Case (50UN) - $29,960.00
    $29,960.00
    Case (50UN)
       $29,960.00
    Supplier Number: JZMZCRMS-10
    Especificaciones:
    Minority or Woman-Owned Business Enterprise
    Small Business Enterprise
      Waste Reducing
    More
    Specifications
    Cat. No.
    89140-706
    Environmentally Preferable
    Waste Reducing
    Minority or Woman-Owned Business Enterprise
    Small Business Enterprise
      Waste Reducing
Accessories
Información del producto
  • Catalog #
    Material
    Length
    Availability
    Price
  • MicroMop™ adapter, attaches to SSU series handles
    MicroMop™ adapter, attaches to SSU series handles
    68400-202
    Stainless steel, 304 electropolished, polypropylene
    -
    Log in Para disponibilidad
    Each (1UN) - $2,540.00
Información del producto
  • Catalog #
    Material
    Length
    Availability
    Price
  • Stainless steel quick disconnect extendable handle
    Stainless steel quick disconnect extendable handle
    68400-164
    Stainless steel 304, electropolished
    Extendable from 96.5 - 182.9 cm (38 - 72")
    Log in Para disponibilidad
    Each (1UN) - $5,810.00