Orders LinkContactUsLinkComponent
VWR® Poly Bags, Pre-Opened
VWR® Poly Bags, Pre-Opened
2 mil., 203×254 mm (8×10")
Catalog # 89071-416
Supplier:   VWR International
VWR® Poly Bags, Pre-Opened
Catalog # 89071-416
Supplier:  VWR International
VWR® Poly Bags, Pre-Opened
VWR® Poly Bags, Pre-Opened
2 mil., 203×254 mm (8×10")
Price
$2,754.60
Log in for availability
Case (1,250UN)
Product Details & Documents
Pre-opened bags on rolls are for use on automated packing machines.
  • Suitable for use with automated packing machines
  • Tubular construction
  • Durable bottom seal
Bags have a slit in front and a perforated back for automatic loading or fast hand loading of products. Made with a tubular construction and a durable bottom seal. Wound on a 8.9 cm (3¹/₂") core. Core plug has a 2.5 cm (1") diameter hole.
Specifications
  • Thickness:
    2 mil.
  • W×L:
    203×254 mm (8×10")