Provide reliable, durable, and comfortable hand protection during demanding cleanroom applications. Textured fingers enhance grip on components.
- Quality tested according to ASTM, FTMS, and IEST standards
- Provide comfort, protection, and tactile sensitivity
- Yield exceptionally low particle counts
- Highly resistant to permeation
- Tapered, beaded cuff provides secure fit
- Recommended for ISO Class 5 (FED-STD-209E Class 100/M3.5) or higher cleanrooms
Manufactured from a synthetic copolymer of acrylonitrile, butadiene, and methylacrylic acid (components that comply with FDA CFR 21). High tensile strength and strong permeation resistance against a wide variety of solvents, acids, and bases. Non-staining, non-smudging gloves reduce the risk of contamination to parts or components being handled by the user, even during prolonged wear.
Static-dissipative to inhibit harmful static discharge. Silicone-free material minimizes ionic contaminants and nonvolatile residue (NVR) transfer. Contain no natural rubber latex. AQL level of 1.5 for pinholes. For single use.
- Catalog No:
- 89130-834
- 89130-836
- 89130-838
- 89130-840
- 89130-842
- 89130-844
- Ambidiestro/Específico para cada mano:
- Ambidiestro
- Ambidiestro
- Ambidiestro
- Ambidiestro
- Ambidiestro
- Ambidiestro
- Con polvo/sin polvo:
- Sin polvo
- Sin polvo
- Sin polvo
- Sin polvo
- Sin polvo
- Sin polvo
- Espesor del puño:
- 0.08–0.12 mm
- 0.08–0.12 mm
- 0.08–0.12 mm
- 0.08–0.12 mm
- 0.08–0.12 mm
- 0.08–0.12 mm
- Espesor del dedo:
- 0.10–0.20 mm
- 0.10–0.20 mm
- 0.10–0.20 mm
- 0.10–0.20 mm
- 0.10–0.20 mm
- 0.10–0.20 mm
- Espesor de la palma:
- 0.10–0.15 mm
- 0.10–0.15 mm
- 0.10–0.15 mm
- 0.10–0.15 mm
- 0.10–0.15 mm
- 0.10–0.15 mm